Search
Search

ALKBH8CACACTTGTTAGATCTGGCCATTAT[C/T]AAAGAGACAAAAGATAACAAACGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ALKBH8 - alkB homolog 8, tRNA methyltransferase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001301010.1 | Intron | NP_001287939.1 | ||||
| NM_138775.2 | Intron | NP_620130.2 | ||||
| XM_005271728.2 | Intron | XP_005271785.1 | ||||
| XM_005271729.3 | Intron | XP_005271786.1 | ||||
| XM_017018553.1 | Intron | XP_016874042.1 | ||||
| XM_017018554.1 | Intron | XP_016874043.1 | ||||
| XM_017018555.1 | Intron | XP_016874044.1 | ||||
| XM_017018556.1 | Intron | XP_016874045.1 | ||||
| XM_017018557.1 | Intron | XP_016874046.1 | ||||