Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTAGCCAGCGCAAAGCATGCG[A/G]GGGGTGAGGGCTCCGCTGTACACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606751 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPRSS5 - transmembrane protease, serine 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288749.1 | 5963 | Silent Mutation | CCC,CCT | P,P 273 | NP_001275678.1 | |
NM_001288750.1 | 5963 | Silent Mutation | CCC,CCT | P,P 342 | NP_001275679.1 | |
NM_001288751.1 | 5963 | Silent Mutation | CCC,CCT | P,P 377 | NP_001275680.1 | |
NM_001288752.1 | 5963 | Silent Mutation | CCC,CCT | P,P 317 | NP_001275681.1 | |
NM_030770.3 | 5963 | Silent Mutation | CCC,CCT | P,P 386 | NP_110397.2 | |
XM_017018366.1 | 5963 | Silent Mutation | CCC,CCT | P,P 156 | XP_016873855.1 | |
XM_017018367.1 | 5963 | Silent Mutation | CCC,CCT | P,P 156 | XP_016873856.1 |