Search
Search

LOC105369535
SORL1CTTAGATTCTCATGGGTGGATATTC[G/T]TTGGAGTTGGTGGGGTTGGAGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369535 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| LOC105369535 - uncharacterized LOC105369535 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| XM_017018646.1 | 2439 | UTR 5 | XP_016874135.1 | |||
| XM_017018647.1 | 2439 | UTR 5 | XP_016874136.1 | |||
| XM_017018648.1 | 2439 | UTR 5 | XP_016874137.1 | |||
| XM_017018649.1 | 2439 | UTR 5 | XP_016874138.1 | |||
| XM_017018650.1 | 2439 | UTR 5 | XP_016874139.1 | |||
| XM_017018651.1 | 2439 | UTR 5 | XP_016874140.1 | |||
| SORL1 - sortilin related receptor 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_003105.5 | 2439 | Intron | NP_003096.1 | |||
| XM_011542963.2 | 2439 | Intron | XP_011541265.1 | |||
| XM_011542965.2 | 2439 | Intron | XP_011541267.1 | |||
| XM_011542967.2 | 2439 | Intron | XP_011541269.1 | |||
| XM_017018169.1 | 2439 | Intron | XP_016873658.1 | |||
| XM_017018170.1 | 2439 | Intron | XP_016873659.1 | |||
| XM_017018171.1 | 2439 | Intron | XP_016873660.1 | |||
| XM_017018172.1 | 2439 | Intron | XP_016873661.1 | |||