Search
Search

C19orf45
PEX11GGTTCCCGCCGCCATCCACGCCGCCG[A/T]GGGAGCTGCTGCAAGCGCAGGCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| C19orf45 - chromosome 19 open reading frame 45 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_198534.2 | 421 | Missense Mutation | AGG,TGG | R,W 94 | NP_940936.2 | |
| XM_005272473.3 | 421 | Missense Mutation | AGG,TGG | R,W 94 | XP_005272530.1 | |
| XM_011527993.1 | 421 | Missense Mutation | AGG,TGG | R,W 94 | XP_011526295.1 | |
| XM_011527995.2 | 421 | Missense Mutation | AGG,TGG | R,W 94 | XP_011526297.1 | |
| PEX11G - peroxisomal biogenesis factor 11 gamma | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||