Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TCCAGGCCAAAGTCCAAATTTGTAC[A/G]TGAGAAAAAAAGGTGTTGAGGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC5D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UNC5D - unc-5 netrin receptor D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322818.1 | Intron | NP_001309747.1 | ||||
NM_080872.3 | Intron | NP_543148.2 | ||||
XM_011544400.2 | Intron | XP_011542702.1 | ||||
XM_011544401.2 | Intron | XP_011542703.1 | ||||
XM_011544402.2 | Intron | XP_011542704.1 | ||||
XM_011544403.2 | Intron | XP_011542705.1 | ||||
XM_011544404.2 | Intron | XP_011542706.1 | ||||
XM_017013044.1 | Intron | XP_016868533.1 | ||||
XM_017013045.1 | Intron | XP_016868534.1 | ||||
XM_017013046.1 | Intron | XP_016868535.1 | ||||
XM_017013047.1 | Intron | XP_016868536.1 | ||||
XM_017013048.1 | Intron | XP_016868537.1 | ||||
XM_017013049.1 | Intron | XP_016868538.1 | ||||
XM_017013050.1 | Intron | XP_016868539.1 |