Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
| Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
| EAS - Not Available | Caucasian
|
YRI (Yoruba) - Not Available | ||||||
| SAS - Not Available | African American
|
JPT (Japanese)
|
||||||
| AFR - Not Available | African American
|
CHB (Han Chinese)
|
||||||
| EUR - Not Available | Japanese
|
|||||||
| AMR - Not Available | Japanese
|
|||||||
Chinese
|
||||||||
Chinese
|
| ABCB1 - ATP binding cassette subfamily B member 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000927.4 | 3170 | Missense Mutation | NP_000918.2 | |||
Set Membership: |
|