Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTCATGGTGTGGTCCCGCGGG[C/T]AGCGGCGCAAGATGGCCCAGGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 184429 MIM: 616338 | ||||||||||||||||||||
Literature Links: |
SOX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SOX2 - SRY-box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003106.3 | 600 | Nonsense Mutation | CAG,TAG | Q,* 55 | NP_003097.1 |
SOX2-OT - SOX2 overlapping transcript | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |