Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGTCAGCTGACTCCACTGTAACACA[C/T]GCCTGCGAAGTATCACGTGATTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BACH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BACH2 - BTB domain and CNC homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170794.1 | 6839 | UTR 3 | NP_001164265.1 | |||
NM_021813.3 | 6839 | UTR 3 | NP_068585.1 | |||
XM_005248758.4 | 6839 | UTR 3 | XP_005248815.1 | |||
XM_005248759.4 | 6839 | UTR 3 | XP_005248816.1 | |||
XM_011536039.2 | 6839 | Intron | XP_011534341.1 | |||
XM_011536040.2 | 6839 | Intron | XP_011534342.1 | |||
XM_017011156.1 | 6839 | UTR 3 | XP_016866645.1 | |||
XM_017011157.1 | 6839 | UTR 3 | XP_016866646.1 | |||
XM_017011158.1 | 6839 | UTR 3 | XP_016866647.1 | |||
XM_017011159.1 | 6839 | UTR 3 | XP_016866648.1 | |||
XM_017011160.1 | 6839 | UTR 3 | XP_016866649.1 | |||
XM_017011161.1 | 6839 | UTR 3 | XP_016866650.1 | |||
XM_017011162.1 | 6839 | UTR 3 | XP_016866651.1 | |||
XM_017011163.1 | 6839 | UTR 3 | XP_016866652.1 | |||
XM_017011164.1 | 6839 | UTR 3 | XP_016866653.1 | |||
XM_017011165.1 | 6839 | Intron | XP_016866654.1 | |||
XM_017011166.1 | 6839 | UTR 3 | XP_016866655.1 | |||
XM_017011167.1 | 6839 | Intron | XP_016866656.1 |