Search
Search

CISD3
MLLT6
PCGF2CCTGGCCTCAAGTTAAGGGGGGCAC[A/G]GGAGCGCCGTTGACAGTCATCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CISD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CISD3 - CDGSH iron sulfur domain 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001136498.1 | 2279 | UTR 3 | NP_001129970.1 | |||
| MLLT6 - MLLT6, PHD finger domain containing | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PCGF2 - polycomb group ring finger 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_007144.2 | 2279 | Silent Mutation | CCC,CCT | P,P 339 | NP_009075.1 | |
| XM_005257640.2 | 2279 | Silent Mutation | CCC,CCT | P,P 339 | XP_005257697.1 | |
| XM_005257641.4 | 2279 | Silent Mutation | CCC,CCT | P,P 339 | XP_005257698.1 | |
| XM_005257642.3 | 2279 | Silent Mutation | CCC,CCT | P,P 339 | XP_005257699.1 | |
| XM_017025016.1 | 2279 | Silent Mutation | CCC,CCT | P,P 339 | XP_016880505.1 | |