Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GCGTGCTGGCCGCCAGCCCGTCCAC[A/G]TCGGGCCCGTCCCCGTCCTCGCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605878 | ||||||||||||||||||||
Literature Links: |
ZBTB7A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB7A - zinc finger and BTB domain containing 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317990.1 | 2131 | Silent Mutation | GAC,GAT | D,D 310 | NP_001304919.1 | |
NM_015898.3 | 2131 | Silent Mutation | GAC,GAT | D,D 310 | NP_056982.1 | |
XM_005259570.4 | 2131 | Silent Mutation | GAC,GAT | D,D 360 | XP_005259627.3 | |
XM_005259571.4 | 2131 | Silent Mutation | GAC,GAT | D,D 310 | XP_005259628.1 |