Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_153441332_10
          See other TMEM189-UBE2V1 GT Assays ›
          SNP ID:
          rs116580959
          Gene
          TMEM189-UBE2V1 UBE2V1
          Gene Name
          TMEM189-UBE2V1 readthrough
          ubiquitin conjugating enzyme E2 V1
          Set Membership:
          -
          Chromosome Location:
          Chr.20: 50082819 - 50082819 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          GCGGCTGAGGGAGTTTCATATTTTC[C/T]TTAGACATCATTAGGCGCCGAAGCT

          Assay ID C_153441332_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 602995

          Literature Links:

          TMEM189-UBE2V1 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          C (0.00)
          (1.00)
          Caucasian - Not Available CEPH (CEU) - Not Available
          EAS
          C (0.01)
          (0.99)
          African American - Not Available YRI (Yoruba) - Not Available
          SAS
          C (0.00)
          (1.00)
          Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR
          C (0.00)
          (1.00)
          Japanese - Not Available JPT (Japanese) - Not Available
          EUR
          C (0.00)
          (1.00)
          AMR
          C (0.00)
          (1.00)
          TMEM189-UBE2V1 - TMEM189-UBE2V1 readthrough
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_199203.2 769 Silent Mutation AAA,AAG K,K 354 NP_954673.1
          UBE2V1 - ubiquitin conjugating enzyme E2 V1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001032288.2 769 Silent Mutation AAA,AAG K,K 131 NP_001027459.1
          NM_001257393.1 769 Silent Mutation AAA,AAG K,K 154 NP_001244322.1
          NM_001257394.1 769 Silent Mutation AAA,AAG K,K 87 NP_001244323.1
          NM_001257395.1 769 Silent Mutation AAA,AAG K,K 128 NP_001244324.1
          NM_001257396.1 769 Silent Mutation AAA,AAG K,K 89 NP_001244325.1
          NM_001257397.1 769 Silent Mutation AAA,AAG K,K 89 NP_001244326.1
          NM_001257398.1 769 Silent Mutation AAA,AAG K,K 89 NP_001244327.1
          NM_001257399.1 769 Silent Mutation AAA,AAG K,K 89 NP_001244328.1
          NM_001282575.2 769 Silent Mutation AAA,AAG K,K 75 NP_001269504.1
          NM_001282576.1 769 Silent Mutation AAA,AAG K,K 89 NP_001269505.1
          NM_001282577.2 769 Silent Mutation AAA,AAG K,K 75 NP_001269506.1
          NM_001282578.2 769 Silent Mutation AAA,AAG K,K 75 NP_001269507.1
          NM_001282579.1 769 Silent Mutation AAA,AAG K,K 89 NP_001269508.1
          NM_001282580.1 769 Intron NP_001269509.1
          NM_021988.5 769 Silent Mutation AAA,AAG K,K 154 NP_068823.2
          NM_022442.5 769 Silent Mutation AAA,AAG K,K 87 NP_071887.1
          NM_199144.2 769 Silent Mutation AAA,AAG K,K 154 NP_954595.1

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          regulation of DNA repair
          postreplication repair
          regulation of transcription, DNA-templated
          cell differentiation
          Fc-epsilon receptor signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          nucleotide-binding oligomerization domain containing signaling pathway
          protein K63-linked ubiquitination
          protein binding
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0