Search
Search

TSPEAR
TSPEAR-AS1
TSPEAR-AS2TCATATGACCAGCACTAGCCAATGA[A/G]CTGTGAGTACACGTGACACAGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPEAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| TSPEAR - thrombospondin type laminin G domain and EAR repeats | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001272037.1 | Intron | NP_001258966.1 | ||||
| NM_144991.2 | Intron | NP_659428.2 | ||||
| TSPEAR-AS1 - TSPEAR antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TSPEAR-AS2 - TSPEAR antisense RNA 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||