Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GGTCCCCAGATTGCAGAGGGAGACG[C/T]GGACGTGAGTGGAGCGGGGCGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610553 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UFM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UFM1 - ubiquitin fold modifier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286703.1 | 53 | UTR 5 | NP_001273632.1 | |||
NM_001286704.1 | 53 | UTR 5 | NP_001273633.1 | |||
NM_001286705.1 | 53 | UTR 5 | NP_001273634.1 | |||
NM_001286706.1 | 53 | UTR 5 | NP_001273635.1 | |||
NM_016617.3 | 53 | UTR 5 | NP_057701.1 |