Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GATGCTGATGCGATATGTAGCCTTT[G/T]ACGTGTCCCTGCGGGGCAGAAAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613713 MIM: 176895 | ||||||||||||||||||||
Literature Links: |
PCID2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCID2 - PCI domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127202.3 | 1712 | Silent Mutation | GTA,GTC | V,V 373 | NP_001120674.1 | |
NM_001127203.3 | 1712 | Silent Mutation | GTA,GTC | V,V 373 | NP_001120675.1 | |
NM_001258213.2 | 1712 | Silent Mutation | GTA,GTC | V,V 371 | NP_001245142.1 | |
NM_001320655.1 | 1712 | Silent Mutation | GTA,GTC | V,V 371 | NP_001307584.1 | |
NM_001320656.1 | 1712 | Silent Mutation | GTA,GTC | V,V 427 | NP_001307585.1 | |
NM_001320657.1 | 1712 | Missense Mutation | AAA,CAA | K,Q 375 | NP_001307586.1 | |
NM_001320659.1 | 1712 | Missense Mutation | AAA,CAA | K,Q 375 | NP_001307588.1 | |
NM_001320660.1 | 1712 | Missense Mutation | AAA,CAA | K,Q 373 | NP_001307589.1 | |
NM_018386.4 | 1712 | Silent Mutation | GTA,GTC | V,V 373 | NP_060856.2 | |
XM_005268325.3 | 1712 | Intron | XP_005268382.1 | |||
XM_017020664.1 | 1712 | Silent Mutation | GTA,GTC | V,V 371 | XP_016876153.1 | |
XM_017020665.1 | 1712 | Silent Mutation | GTA,GTC | V,V 331 | XP_016876154.1 | |
XM_017020666.1 | 1712 | Silent Mutation | GTA,GTC | V,V 331 | XP_016876155.1 | |
XM_017020667.1 | 1712 | Silent Mutation | GTA,GTC | V,V 206 | XP_016876156.1 | |
XM_017020668.1 | 1712 | Silent Mutation | GTA,GTC | V,V 206 | XP_016876157.1 | |
XM_017020669.1 | 1712 | Silent Mutation | GTA,GTC | V,V 206 | XP_016876158.1 | |
XM_017020670.1 | 1712 | Silent Mutation | GTA,GTC | V,V 206 | XP_016876159.1 | |
XM_017020671.1 | 1712 | Silent Mutation | GTA,GTC | V,V 206 | XP_016876160.1 | |
XM_017020672.1 | 1712 | Silent Mutation | GTA,GTC | V,V 206 | XP_016876161.1 |
PROZ - protein Z, vitamin K dependent plasma glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |