Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GCCCAGTCAATCAAAGGTAGGAGAA[A/G]TGGCTTCTTTCTATACTCAGACTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186790 MIM: 186740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD3D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD3D - CD3d molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD3G - CD3g molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000073.2 | Intron | NP_000064.1 | ||||
XM_005271724.3 | Intron | XP_005271781.1 | ||||
XM_006718941.2 | Intron | XP_006719004.1 |