Search
Search

ESAM
LOC101929340
MSANTD2GTTCGCTATATATCTTGCTTGCTCA[A/C]AATGAGCATTTTCAGGTGAGGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ESAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ESAM - endothelial cell adhesion molecule | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC101929340 - uncharacterized LOC101929340 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MSANTD2 - Myb/SANT DNA binding domain containing 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001308027.1 | 3270 | UTR 3 | NP_001294956.1 | |||
| NM_001312919.1 | 3270 | UTR 3 | NP_001299848.1 | |||
| NM_001312920.1 | 3270 | UTR 3 | NP_001299849.1 | |||
| NM_001312921.1 | 3270 | UTR 3 | NP_001299850.1 | |||
| NM_024631.3 | 3270 | UTR 3 | NP_078907.2 | |||
| XM_005271674.3 | 3270 | Intron | XP_005271731.3 | |||
| XM_006718905.1 | 3270 | UTR 3 | XP_006718968.1 | |||
| XM_006718906.3 | 3270 | UTR 3 | XP_006718969.1 | |||
| XM_011542981.2 | 3270 | Intron | XP_011541283.2 | |||
| XM_011542982.2 | 3270 | Intron | XP_011541284.2 | |||
| XM_017018295.1 | 3270 | UTR 3 | XP_016873784.1 | |||
| XM_017018296.1 | 3270 | UTR 3 | XP_016873785.1 | |||
| XM_017018297.1 | 3270 | UTR 3 | XP_016873786.1 | |||
| XM_017018298.1 | 3270 | UTR 3 | XP_016873787.1 | |||
| XM_017018299.1 | 3270 | Intron | XP_016873788.1 | |||
| XM_017018300.1 | 3270 | Intron | XP_016873789.1 | |||
| XM_017018301.1 | 3270 | Intron | XP_016873790.1 | |||