Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCCTTTCTTCTATCAGGCGAGTCAA[A/G]TAATCATCCAAACCTTCTTCCAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613446 | ||||||||||||||||||||
Literature Links: |
CEP120 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP120 - centrosomal protein 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166226.1 | 3282 | Silent Mutation | TAC,TAT | Y,Y 918 | NP_001159698.1 | |
NM_153223.3 | 3282 | Silent Mutation | TAC,TAT | Y,Y 944 | NP_694955.2 | |
XM_005271901.4 | 3282 | Silent Mutation | TAC,TAT | Y,Y 899 | XP_005271958.1 | |
XM_011543185.2 | 3282 | Silent Mutation | TAC,TAT | Y,Y 918 | XP_011541487.1 | |
XM_011543186.2 | 3282 | Silent Mutation | TAC,TAT | Y,Y 455 | XP_011541488.1 | |
XM_017009085.1 | 3282 | Silent Mutation | TAC,TAT | Y,Y 455 | XP_016864574.1 |