Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAAGAAAGGCACCTGGAGGCC[A/G]AAGGTGCTCTGCGGGTGAAGAGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613104 MIM: 606813 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACFD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACFD1 - calcium channel flower domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC2A6 - solute carrier family 2 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145099.1 | 1444 | Silent Mutation | TTC,TTT | F,F 397 | NP_001138571.1 | |
NM_017585.3 | 1444 | Silent Mutation | TTC,TTT | F,F 459 | NP_060055.2 | |
XM_011518189.2 | 1444 | Silent Mutation | TTC,TTT | F,F 253 | XP_011516491.2 | |
XM_017014236.1 | 1444 | Intron | XP_016869725.1 | |||
XM_017014237.1 | 1444 | Silent Mutation | TTC,TTT | F,F 215 | XP_016869726.1 | |
XM_017014238.1 | 1444 | Silent Mutation | TTC,TTT | F,F 215 | XP_016869727.1 |