Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GTCTTTTAAACAATTCGACAATAAA[A/G]AAGTACAATTTTTTTTGTGCTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPC1 - enhancer of polycomb homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272004.1 | 3743 | UTR 3 | NP_001258933.1 | |||
NM_001272019.2 | 3743 | UTR 3 | NP_001258948.1 | |||
NM_001282391.1 | 3743 | UTR 3 | NP_001269320.1 | |||
NM_025209.3 | 3743 | UTR 3 | NP_079485.1 | |||
XM_006717510.1 | 3743 | UTR 3 | XP_006717573.1 |