Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
TGACTGTTTCTTTGTGCATCTTCAA[C/T]AGGGTATGGAGAGCGGGGAGCTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604189 MIM: 186946 MIM: 600230 MIM: 601140 MIM: 601398 | ||||||||||||||||||||
Literature Links: |
DNAJC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC4 - DnaJ heat shock protein family (Hsp40) member C4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FKBP2 - FK506 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135208.1 | Intron | NP_001128680.1 | ||||
NM_004470.3 | Intron | NP_004461.2 | ||||
NM_057092.2 | Intron | NP_476433.1 | ||||
XM_005273848.3 | Intron | XP_005273905.1 |
LOC105369340 - uncharacterized LOC105369340 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCB3 - phospholipase C beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R14B - protein phosphatase 1 regulatory inhibitor subunit 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138689.2 | Intron | NP_619634.1 |
VEGFB - vascular endothelial growth factor B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |