Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GTTTTTGCTCTTGGTAATTGTCTCT[C/T]GGCCCTTGGTGGGAACGCCATCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF22 - immunoglobulin superfamily member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173588.3 | 4151 | Missense Mutation | CAA,CGA | Q,R 1048 | NP_775859.3 | |
XM_011520030.2 | 4151 | Missense Mutation | CAA,CGA | Q,R 1068 | XP_011518332.1 | |
XM_011520031.2 | 4151 | Missense Mutation | CAA,CGA | Q,R 967 | XP_011518333.1 | |
XM_017017607.1 | 4151 | Missense Mutation | CAA,CGA | Q,R 1068 | XP_016873096.1 | |
XM_017017608.1 | 4151 | Missense Mutation | CAA,CGA | Q,R 1048 | XP_016873097.1 |
TMEM86A - transmembrane protein 86A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |