Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CACACACCAGGATGAGCGTGGACTT[C/T]GCTGGAGGAGGAAGGTGCTCCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608342 MIM: 603142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYB5R2 - cytochrome b5 reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302826.1 | 946 | Silent Mutation | NP_001289755.1 | |||
NM_001302827.1 | 946 | UTR 3 | NP_001289756.1 | |||
NM_016229.4 | 946 | Silent Mutation | NP_057313.2 | |||
XM_005252975.4 | 946 | Silent Mutation | XP_005253032.1 | |||
XM_006718251.2 | 946 | Silent Mutation | XP_006718314.1 | |||
XM_011520184.2 | 946 | Silent Mutation | XP_011518486.1 | |||
XM_011520185.2 | 946 | UTR 3 | XP_011518487.1 | |||
XM_017017920.1 | 946 | Silent Mutation | XP_016873409.1 | |||
XM_017017921.1 | 946 | UTR 3 | XP_016873410.1 | |||
XM_017017922.1 | 946 | UTR 3 | XP_016873411.1 |