Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAAACATACAGTTTGAACCAAG[C/T]ATATGTGATGCCTACTGCACAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG8 - ALG8, alpha-1,3-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007027.2 | 1581 | UTR 3 | NP_001007028.1 | |||
NM_024079.4 | 1581 | Missense Mutation | ACT,GCT | T,A 506 | NP_076984.2 | |
XM_005274247.3 | 1581 | Missense Mutation | ACT,GCT | T,A 497 | XP_005274304.1 | |
XM_011545252.2 | 1581 | Missense Mutation | ACT,GCT | T,A 418 | XP_011543554.1 | |
XM_017018274.1 | 1581 | UTR 3 | XP_016873763.1 |