Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTTTTAAATCCTTTTGAAAGT[C/T]CTAGTGATTATTCTAATCTCCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610510 MIM: 613448 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BORA - bora, aurora kinase A activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286746.1 | 356 | Missense Mutation | CCT,TCT | P,S 103 | NP_001273675.1 | |
NM_001286747.1 | 356 | UTR 5 | NP_001273676.1 | |||
NM_024808.3 | 356 | Missense Mutation | CCT,TCT | P,S 88 | NP_079084.3 | |
XM_006719868.3 | 356 | Missense Mutation | CCT,TCT | P,S 88 | XP_006719931.2 |
MZT1 - mitotic spindle organizing protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |