Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600292 MIM: 604441 MIM: 601531 MIM: 605773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY4 - adenylate cyclase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIDEB - cell death-inducing DFFA-like effector b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318807.1 | 45 | Intron | NP_001305736.1 | |||
NM_014430.2 | 45 | Intron | NP_055245.2 | |||
XM_011536659.2 | 45 | Intron | XP_011534961.1 | |||
XM_017021221.1 | 45 | Intron | XP_016876710.1 |
LTB4R - leukotriene B4 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143919.2 | 45 | UTR 5 | NP_001137391.1 | |||
NM_181657.3 | 45 | Intron | NP_858043.1 |
LTB4R2 - leukotriene B4 receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164692.2 | 45 | Silent Mutation | GGA,GGC | G,G 293 | NP_001158164.1 | |
NM_019839.4 | 45 | Silent Mutation | GGA,GGC | G,G 293 | NP_062813.2 |
NOP9 - NOP9 nucleolar protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |