Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGGGTAAACCAGACTGCCCATT[C/T]GACTTGTTCCTTTGAAGATGACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601930 | ||||||||||||||||||||
Literature Links: |
BNC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BNC1 - basonuclin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301206.1 | 2701 | Missense Mutation | CAA,CGA | Q,R 819 | NP_001288135.1 | |
NM_001717.3 | 2701 | Missense Mutation | CAA,CGA | Q,R 826 | NP_001708.3 | |
XM_011521893.1 | 2701 | Missense Mutation | CAA,CGA | Q,R 801 | XP_011520195.1 | |
XM_011521894.1 | 2701 | Missense Mutation | CAA,CGA | Q,R 708 | XP_011520196.1 |