Search Thermo Fisher Scientific
AGAATGGGGACAGGGGGCTGCCATC[A/G]TGCAGCAGAGACTGGGCACTCAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 177070 MIM: 603805 | ||||||||||||||||||||
Literature Links: |
EPB42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPB42 - erythrocyte membrane protein band 4.2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGM5 - transglutaminase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004245.3 | 864 | Silent Mutation | CAC,CAT | H,H 468 | NP_004236.1 | |
NM_201631.3 | 864 | Silent Mutation | CAC,CAT | H,H 550 | NP_963925.2 | |
XM_011522230.2 | 864 | Silent Mutation | CAC,CAT | H,H 207 | XP_011520532.1 | |
XM_017022729.1 | 864 | Silent Mutation | CAC,CAT | H,H 207 | XP_016878218.1 |