Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TCAACCATGAGGTCCACAGTCTCCT[A/C]TGGCAGGGAGCGCCCCACAAACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613374 MIM: 601292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGF29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGF29 - SAGA complex associated factor 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138414.2 | 884 | Intron | NP_612423.1 | |||
XM_017022894.1 | 884 | Intron | XP_016878383.1 |
SULT1A2 - sulfotransferase family 1A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001054.3 | 884 | Nonsense Mutation | GAG,TAG | E,* 217 | NP_001045.1 | |
NM_177528.2 | 884 | Nonsense Mutation | GAG,TAG | E,* 217 | NP_803564.1 | |
XM_006721077.3 | 884 | Nonsense Mutation | GAG,TAG | E,* 184 | XP_006721140.1 | |
XM_017023600.1 | 884 | Nonsense Mutation | GAG,TAG | E,* 223 | XP_016879089.1 | |
XM_017023601.1 | 884 | Nonsense Mutation | GAG,TAG | E,* 223 | XP_016879090.1 | |
XM_017023602.1 | 884 | Nonsense Mutation | GAG,TAG | E,* 223 | XP_016879091.1 | |
XM_017023603.1 | 884 | Nonsense Mutation | GAG,TAG | E,* 143 | XP_016879092.1 |