Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108745 MIM: 611113 MIM: 605213 | ||||||||||||||||||||
Literature Links: |
AMDHD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMDHD2 - amidohydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145815.1 | 780 | Missense Mutation | ACC,ATC | T,I 228 | NP_001139287.1 | |
NM_015944.3 | 780 | Missense Mutation | ACC,ATC | T,I 228 | NP_057028.2 | |
XM_017023262.1 | 780 | Missense Mutation | ACC,ATC | T,I 228 | XP_016878751.1 | |
XM_017023263.1 | 780 | Missense Mutation | ACC,ATC | T,I 228 | XP_016878752.1 | |
XM_017023264.1 | 780 | Missense Mutation | ACC,ATC | T,I 228 | XP_016878753.1 | |
XM_017023265.1 | 780 | Missense Mutation | ACC,ATC | T,I 228 | XP_016878754.1 | |
XM_017023266.1 | 780 | Missense Mutation | ACC,ATC | T,I 65 | XP_016878755.1 | |
XM_017023267.1 | 780 | UTR 5 | XP_016878756.1 |
ATP6V0C - ATPase H+ transporting V0 subunit c | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CEMP1 - cementum protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3178 - microRNA 3178 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDPK1 - 3-phosphoinositide dependent protein kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |