Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGCTGGCCCTTCAGCTCCGAGCACC[A/G]CTCCTGAAGCTTCCGCTCCGACTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613479 | ||||||||||||||||||||
Literature Links: |
AATK-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AATK-AS1 - AATK antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CEP131 - centrosomal protein 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009811.3 | 3092 | Missense Mutation | CGG,TGG | R,W 911 | NP_001009811.2 | |
NM_001319228.1 | 3092 | Missense Mutation | CGG,TGG | R,W 950 | NP_001306157.1 | |
NM_001319229.1 | 3092 | Missense Mutation | CGG,TGG | R,W 914 | NP_001306158.1 | |
NM_014984.3 | 3092 | Missense Mutation | CGG,TGG | R,W 947 | NP_055799.2 | |
XM_011524547.2 | 3092 | Missense Mutation | CGG,TGG | R,W 955 | XP_011522849.1 | |
XM_011524548.2 | 3092 | Missense Mutation | CGG,TGG | R,W 952 | XP_011522850.1 | |
XM_011524550.2 | 3092 | Missense Mutation | CGG,TGG | R,W 949 | XP_011522852.1 | |
XM_011524551.2 | 3092 | Missense Mutation | CGG,TGG | R,W 955 | XP_011522853.1 | |
XM_011524552.2 | 3092 | Missense Mutation | CGG,TGG | R,W 919 | XP_011522854.1 | |
XM_011524553.2 | 3092 | Missense Mutation | CGG,TGG | R,W 914 | XP_011522855.1 | |
XM_011524555.2 | 3092 | Missense Mutation | CGG,TGG | R,W 955 | XP_011522857.1 | |
XM_017024369.1 | 3092 | Missense Mutation | CGG,TGG | R,W 950 | XP_016879858.1 |