Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGCCGATGGCGGCAGCAGCTGC[A/G]GCAGCATGATGTTGTAGGATGGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614396 MIM: 607759 | ||||||||||||||||||||
Literature Links: |
GPATCH8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPATCH8 - G-patch domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002909.3 | 4254 | Silent Mutation | GCC,GCT | A,A 1385 | NP_001002909.1 | |
NM_001304939.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1360 | NP_001291868.1 | |
NM_001304940.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | NP_001291869.1 | |
NM_001304941.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | NP_001291870.1 | |
NM_001304942.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | NP_001291871.1 | |
NM_001304943.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | NP_001291872.1 | |
XM_011524557.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1414 | XP_011522859.1 | |
XM_011524558.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1336 | XP_011522860.1 | |
XM_011524559.2 | 4254 | Silent Mutation | GCC,GCT | A,A 1336 | XP_011522861.1 | |
XM_011524561.2 | 4254 | Silent Mutation | GCC,GCT | A,A 1336 | XP_011522863.1 | |
XM_017024372.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1336 | XP_016879861.1 | |
XM_017024373.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1336 | XP_016879862.1 | |
XM_017024374.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1336 | XP_016879863.1 | |
XM_017024375.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | XP_016879864.1 | |
XM_017024376.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | XP_016879865.1 | |
XM_017024377.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | XP_016879866.1 | |
XM_017024378.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | XP_016879867.1 | |
XM_017024379.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1307 | XP_016879868.1 | |
XM_017024380.1 | 4254 | Silent Mutation | GCC,GCT | A,A 1171 | XP_016879869.1 |
ITGA2B - integrin subunit alpha 2b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |