Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
TGTCGTGAAAGCGAACGAAGGCGAA[A/G]CCGCGGGACTCCTTGGTGTAGCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615262 MIM: 600813 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL23 - methyltransferase like 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR636 - microRNA 636 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRSF2 - serine and arginine rich splicing factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195427.1 | 419 | Silent Mutation | GGC,GGT | G,G 56 | NP_001182356.1 | |
NM_003016.4 | 419 | Silent Mutation | GGC,GGT | G,G 56 | NP_003007.2 | |
XM_017024942.1 | 419 | Silent Mutation | GGC,GGT | G,G 56 | XP_016880431.1 |