Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CACTGCTCCTAGCCCCATCTTACCC[A/G]AAAAGTCTTGCCAGCTACTTAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604399 MIM: 607048 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R1B - protein phosphatase 1 regulatory inhibitor subunit 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STARD3 - StAR related lipid transfer domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165937.1 | Intron | NP_001159409.1 | ||||
NM_001165938.1 | Intron | NP_001159410.1 | ||||
NM_006804.3 | Intron | NP_006795.3 | ||||
XM_006721646.2 | Intron | XP_006721709.1 | ||||
XM_011524210.1 | Intron | XP_011522512.1 | ||||
XM_011524211.2 | Intron | XP_011522513.1 | ||||
XM_011524213.1 | Intron | XP_011522515.1 | ||||
XM_017024041.1 | Intron | XP_016879530.1 |