Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CACCTTCCAGCCTCTGGCTACAACA[C/T]GCCTGCAGCCCAAGGCAAAAGCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610520 | ||||||||||||||||||||
Literature Links: |
CD300LG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD300LG - CD300 molecule like family member g | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168322.1 | 873 | Missense Mutation | CGC,TGC | R,C 145 | NP_001161794.1 | |
NM_001168323.1 | 873 | Intron | NP_001161795.1 | |||
NM_001168324.1 | 873 | Intron | NP_001161796.1 | |||
NM_145273.3 | 873 | Missense Mutation | CGC,TGC | R,C 145 | NP_660316.2 | |
XM_005257075.2 | 873 | Missense Mutation | CGC,TGC | R,C 145 | XP_005257132.1 | |
XM_005257076.2 | 873 | Intron | XP_005257133.1 | |||
XM_006721721.2 | 873 | Missense Mutation | CGC,TGC | R,C 145 | XP_006721784.1 | |
XM_006721722.2 | 873 | Intron | XP_006721785.1 | |||
XM_011524384.2 | 873 | Missense Mutation | CGC,TGC | R,C 145 | XP_011522686.1 | |
XM_017024235.1 | 873 | Intron | XP_016879724.1 |