Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CAGTCTCAAAGCCAAAAGAGTCGAT[A/G]GCCGGGATAAAAGCTTTGATGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603892 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFTUD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFTUD2 - elongation factor Tu GTP binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142605.1 | 3029 | Silent Mutation | GCC,GCT | A,A 842 | NP_001136077.1 | |
NM_001258353.1 | 3029 | Silent Mutation | GCC,GCT | A,A 877 | NP_001245282.1 | |
NM_001258354.1 | 3029 | Silent Mutation | GCC,GCT | A,A 867 | NP_001245283.1 | |
NM_004247.3 | 3029 | Silent Mutation | GCC,GCT | A,A 877 | NP_004238.3 |
HIGD1B - HIG1 hypoxia inducible domain family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |