Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTACATACCAGGATAGCCACCAA[C/T]ATAAATGGGATTGTTGGTGTCCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 150320 | ||||||||||||||||||||
Literature Links: |
LAMA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAMA1 - laminin subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005559.3 | 9146 | Missense Mutation | ATT,GTT | I,V 3018 | NP_005550.2 | |
XM_011525655.2 | 9146 | Missense Mutation | ATT,GTT | I,V 3028 | XP_011523957.1 | |
XM_011525656.2 | 9146 | Missense Mutation | ATT,GTT | I,V 2504 | XP_011523958.1 | |
XM_011525657.1 | 9146 | Intron | XP_011523959.1 |