Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
CGCAGGGTCCTTGGCATCAGCCCTC[C/T]CTGGGTGTTTCCATTTATCCTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609150 MIM: 156535 | ||||||||||||||||||||
Literature Links: |
CXXC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CXXC1 - CXXC finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101654.1 | 1563 | Missense Mutation | GAG,GGG | E,G 366 | NP_001095124.1 | |
NM_014593.3 | 1563 | Missense Mutation | GAG,GGG | E,G 362 | NP_055408.2 | |
XM_011525940.2 | 1563 | Missense Mutation | GAG,GGG | E,G 366 | XP_011524242.1 | |
XM_011525941.2 | 1563 | Missense Mutation | GAG,GGG | E,G 241 | XP_011524243.1 | |
XM_017025718.1 | 1563 | Missense Mutation | GAG,GGG | E,G 362 | XP_016881207.1 | |
XM_017025719.1 | 1563 | Missense Mutation | GAG,GGG | E,G 237 | XP_016881208.1 |
MBD1 - methyl-CpG binding domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |