Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
TGTAAAATAAGGGCTGAGCTCTGAC[C/T]AAAGATTTTGCCACATTCATCACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609600 | ||||||||||||||||||||
Literature Links: |
ZNF396 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF396 - zinc finger protein 396 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322286.1 | 913 | Missense Mutation | AGT,GGT | S,G 261 | NP_001309215.1 | |
NM_001322290.1 | 913 | Missense Mutation | AGT,GGT | S,G 261 | NP_001309219.1 | |
NM_001322291.1 | 913 | Intron | NP_001309220.1 | |||
NM_145756.2 | 913 | Missense Mutation | AGT,GGT | S,G 261 | NP_665699.1 | |
XM_006722432.3 | 913 | Missense Mutation | AGT,GGT | S,G 261 | XP_006722495.1 | |
XM_017025687.1 | 913 | Missense Mutation | AGT,GGT | S,G 261 | XP_016881176.1 | |
XM_017025688.1 | 913 | Missense Mutation | AGT,GGT | S,G 261 | XP_016881177.1 | |
XM_017025689.1 | 913 | Intron | XP_016881178.1 |