Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CTCGCCAACTGCCCAGGGGCCGCCA[C/T]GTGGGGCAGGCCAGCTGGTGGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611951 MIM: 190180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B9D2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B9D2 - B9 protein domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030578.3 | 556 | Silent Mutation | ACA,ACG | T,T 112 | NP_085055.2 | |
XM_011527349.2 | 556 | Silent Mutation | ACA,ACG | T,T 112 | XP_011525651.1 | |
XM_011527350.1 | 556 | Silent Mutation | ACA,ACG | T,T 59 | XP_011525652.1 |
TGFB1 - transforming growth factor beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527242.1 | 556 | Intron | XP_011525544.1 |
TMEM91 - transmembrane protein 91 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |