Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
CTTTCTCCAGTGTGAGTTCGTTGGT[A/G]CAGAACAAGTGTGTGTTTCCGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF671 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF671 - zinc finger protein 671 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321375.1 | 1835 | Missense Mutation | CAC,TAC | H,Y 317 | NP_001308304.1 | |
NM_001321376.1 | 1835 | Missense Mutation | CAC,TAC | H,Y 338 | NP_001308305.1 | |
NM_024833.2 | 1835 | Missense Mutation | CAC,TAC | H,Y 415 | NP_079109.2 | |
XM_017027314.1 | 1835 | Missense Mutation | CAC,TAC | H,Y 338 | XP_016882803.1 |