Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CTCTTCCTGCTTCCGGTGGGCTCCT[C/T]GGAAGGCCAGGACCAGGGCGCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 132880 MIM: 611810 | ||||||||||||||||||||
Literature Links: |
NR2F6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR2F6 - nuclear receptor subfamily 2 group F member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USHBP1 - USH1 protein network component harmonin binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297703.1 | 1968 | Missense Mutation | CAA,CGA | Q,R 584 | NP_001284632.1 | |
NM_001321417.1 | 1968 | Missense Mutation | CAA,CGA | Q,R 648 | NP_001308346.1 | |
NM_031941.3 | 1968 | Missense Mutation | CAA,CGA | Q,R 648 | NP_114147.2 | |
XM_011528351.2 | 1968 | Intron | XP_011526653.1 |