Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_165628104_10
          See other MASP2 GT Assays ›
          SNP ID:
          rs80356730
          Gene
          MASP2 TARDBP
          Gene Name
          mannan binding lectin serine peptidase 2
          TAR DNA binding protein
          Set Membership:
          -
          Chromosome Location:
          Chr.1: 11022418 - 11022418 on Build GRCh38
          Polymorphism:
          A/G, Transition substitution
          Context Sequence [VIC/FAM]:

          AGCACTACAGAGCAGTTGGGGTATG[A/G]TGGGCATGTTAGCCAGCCAGCAGAA

          Assay ID C_165628104_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          3 submissions

          Phenotype:

          MIM: 605102 MIM: 605078

          Literature Links:

          MASP2 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          MASP2 - mannan binding lectin serine peptidase 2
          There are no transcripts associated with this gene.
          TARDBP - TAR DNA binding protein
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_007375.3 1131 Missense Mutation ATG,GTG M,V 337 NP_031401.1
          XM_017000863.1 1131 Missense Mutation ATG,GTG M,V 337 XP_016856352.1
          XM_017000864.1 1131 Missense Mutation ATG,GTG M,V 337 XP_016856353.1
          XM_017000865.1 1131 Missense Mutation ATG,GTG M,V 337 XP_016856354.1
          XM_017000866.1 1131 Missense Mutation ATG,GTG M,V 337 XP_016856355.1
          XM_017000867.1 1131 Missense Mutation ATG,GTG M,V 337 XP_016856356.1
          XM_017000868.1 1131 Missense Mutation ATG,GTG M,V 337 XP_016856357.1

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          transcription from RNA polymerase II promoter
          mRNA processing
          RNA splicing
          negative regulation of gene expression
          nuclear fragmentation involved in apoptotic nuclear change
          positive regulation of insulin secretion
          response to endoplasmic reticulum stress
          negative regulation by host of viral transcription
          positive regulation of transcription from RNA polymerase II promoter
          regulation of cell cycle
          3'-UTR-mediated mRNA stabilization
          nuclear inner membrane organization
          nucleotide binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline