Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGCCTTTATAGACACTGAAACA[C/T]TGGTGTTCTGTGGTCCTGATTGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPZA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPZA1 - capping actin protein of muscle Z-line alpha subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006135.2 | Intron | NP_006126.1 | ||||
XM_011542225.2 | Intron | XP_011540527.1 | ||||
XM_017002424.1 | Intron | XP_016857913.1 |
ST7L - suppression of tumorigenicity 7 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |