Search
Search

LCE2A
LCE4AGTTGCTGCAGCTCTGAGGGAGGTGG[C/T]TGCTGCCTGAGCCACCACAGACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
LCE2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| LCE2A - late cornified envelope 2A | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LCE4A - late cornified envelope 4A | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_178356.3 | Intron | NP_848133.1 | ||||