Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
AGGCTCCTGCCCCCATGAAGACACC[C/T]GAGGCTGGCTTGGCGGGGAGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBLIM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBLIM1 - filamin binding LIM protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024215.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | NP_001019386.1 | |
NM_001024216.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | NP_001019387.1 | |
NM_017556.2 | 1085 | Silent Mutation | NP_060026.2 | |||
XM_005245900.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_005245957.1 | |
XM_005245901.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_005245958.1 | |
XM_005245902.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_005245959.1 | |
XM_005245903.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_005245960.1 | |
XM_005245909.3 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_005245966.1 | |
XM_006710704.3 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_006710767.1 | |
XM_006710705.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_006710768.1 | |
XM_011541616.2 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_011539918.1 | |
XM_011541617.2 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_011539919.1 | |
XM_011541618.2 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_011539920.1 | |
XM_017001519.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857008.1 | |
XM_017001520.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857009.1 | |
XM_017001521.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857010.1 | |
XM_017001522.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857011.1 | |
XM_017001523.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857012.1 | |
XM_017001524.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857013.1 | |
XM_017001525.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857014.1 | |
XM_017001526.1 | 1085 | Silent Mutation | CCC,CCT | P,P 52 | XP_016857015.1 |