Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
CGGCAGCTTCGTAGATGGGCATGTC[A/G]TTGTGGCGGATGTACTCAGCGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBR4 - ubiquitin protein ligase E3 component n-recognin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020765.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5130 | NP_065816.2 | |
XM_011541108.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5181 | XP_011539410.1 | |
XM_011541109.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5180 | XP_011539411.1 | |
XM_011541110.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5180 | XP_011539412.1 | |
XM_011541111.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5180 | XP_011539413.1 | |
XM_011541112.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5176 | XP_011539414.1 | |
XM_011541113.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5175 | XP_011539415.1 | |
XM_011541114.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5175 | XP_011539416.1 | |
XM_011541115.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5173 | XP_011539417.1 | |
XM_011541116.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5170 | XP_011539418.1 | |
XM_011541117.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5153 | XP_011539419.1 | |
XM_011541118.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5152 | XP_011539420.1 | |
XM_011541119.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5141 | XP_011539421.1 | |
XM_011541120.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5140 | XP_011539422.1 | |
XM_011541121.2 | 15371 | Silent Mutation | AAC,AAT | N,N 5129 | XP_011539423.1 | |
XM_017000822.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5174 | XP_016856311.1 | |
XM_017000823.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5165 | XP_016856312.1 | |
XM_017000824.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5147 | XP_016856313.1 | |
XM_017000825.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5142 | XP_016856314.1 | |
XM_017000826.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5141 | XP_016856315.1 | |
XM_017000827.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5136 | XP_016856316.1 | |
XM_017000828.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5128 | XP_016856317.1 | |
XM_017000829.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5112 | XP_016856318.1 | |
XM_017000830.1 | 15371 | Silent Mutation | AAC,AAT | N,N 5095 | XP_016856319.1 |