Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TCCCTTCCTGGGTGCATTGGTTTGG[A/T]TGCAGCAACAGCTACAGTGGAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604396 | ||||||||||||||||||||
Literature Links: |
SETDB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SETDB1 - SET domain bifurcated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145415.1 | 222 | Missense Mutation | GAT,GTT | D,V 11 | NP_001138887.1 | |
NM_001243491.1 | 222 | Missense Mutation | GAT,GTT | D,V 11 | NP_001230420.1 | |
NM_012432.3 | 222 | Missense Mutation | GAT,GTT | D,V 11 | NP_036564.3 | |
XM_005245641.3 | 222 | Missense Mutation | GAT,GTT | D,V 11 | XP_005245698.1 | |
XM_017002953.1 | 222 | Missense Mutation | GAT,GTT | D,V 11 | XP_016858442.1 | |
XM_017002954.1 | 222 | Missense Mutation | GAT,GTT | D,V 11 | XP_016858443.1 | |
XM_017002955.1 | 222 | Missense Mutation | GAT,GTT | D,V 11 | XP_016858444.1 |