Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CTACGCCAACTTCTTTCCCTATGGT[C/G]ACGCCTCCAAGTTTGCCGAGCACGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 142622 | ||||||||||||||||||||
Literature Links: |
HPCA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HPCA - hippocalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002143.2 | 690 | Missense Mutation | CAC,GAC | H,D 60 | NP_002134.2 | |
XM_005270792.2 | 690 | Missense Mutation | CAC,GAC | H,D 60 | XP_005270849.1 | |
XM_017001118.1 | 690 | Missense Mutation | CAC,GAC | H,D 60 | XP_016856607.1 |
TMEM54 - transmembrane protein 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |