Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCACGCATGCGCGCGGCTCGCC[A/G]CGGTCTTCACTGCGCAGGCGCCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMJD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JMJD4 - jumonji domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161465.1 | 431 | Intron | NP_001154937.1 | |||
NM_023007.2 | 431 | Intron | NP_075383.2 | |||
XM_011544262.2 | 431 | Intron | XP_011542564.1 |
SNAP47 - synaptosome associated protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323930.1 | 431 | Intron | NP_001310859.1 | |||
NM_001323931.1 | 431 | UTR 5 | NP_001310860.1 | |||
NM_001323932.1 | 431 | Intron | NP_001310861.1 | |||
NM_001323933.1 | 431 | Intron | NP_001310862.1 | |||
NM_001323934.1 | 431 | Intron | NP_001310863.1 | |||
NM_001323935.1 | 431 | Missense Mutation | CAC,CGC | H,R 6 | NP_001310864.1 | |
NM_053052.3 | 431 | Missense Mutation | CAC,CGC | H,R 6 | NP_444280.2 | |
XM_006711734.3 | 431 | Intron | XP_006711797.1 | |||
XM_017000231.1 | 431 | Missense Mutation | CAC,CGC | H,R 6 | XP_016855720.1 | |
XM_017000232.1 | 431 | Missense Mutation | CAC,CGC | H,R 6 | XP_016855721.1 |