Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTGGGACCCAGCCCGCAACGAC[A/T]CGCTGCCGCCCACGCTGACCCCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604658 | ||||||||||||||||||||
Literature Links: |
GPR137B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR137B - G protein-coupled receptor 137B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003272.3 | 173 | Missense Mutation | ACG,TCG | T,S 28 | NP_003263.1 | |
XM_017002209.1 | 173 | Missense Mutation | ACG,TCG | T,S 28 | XP_016857698.1 | |
XM_017002210.1 | 173 | Missense Mutation | ACG,TCG | T,S 28 | XP_016857699.1 |